Learn More
Gardner (ASTM D1544, D6166) Colour Reference Standard, Paragon Scientific™
Gardner Colour Reference Standards are for routine calibration of colour measuring instruments and verification of test data. Process industries throughout the world use colorimeters for colour control of products such as edible and industrial oils, fuels, chemicals, pharmaceuticals, paints and coatings.
Brand: Paragon Scientific 134210
Additional Details : CAS Number : 8042-47-5 Weight : 0.90000kg
Description
- Certified according to ISO 17025 / ISO 17034 under UKAS accreditation
- Full Traceability to International Standards
- 500mL Pack Size supplied with MSDS
- 18 months shelf life
- Tamper evident security packaging
- Manufactured in the United kingdom
Specifications
500 mL | |
Color Reference Standard |
Safety and Handling
- GARDNER COLOUR STANDARDS
Signal Word
- Danger
Hazard Category
- Aspiration toxicity Category 1
Hazard Statement
- H304-May be fatal if swallowed and enters airways.
Precautionary Statement
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
Supplemental information
- MIXTURE LIST-Contains : white oil 2
Nominal values quoted