missing translation for 'onlineSavingsMsg'
Learn More

Gardner (ASTM D1544, D6166) Colour Reference Standard, Paragon Scientific™

Gardner Colour Reference Standards are for routine calibration of colour measuring instruments and verification of test data. Process industries throughout the world use colorimeters for colour control of products such as edible and industrial oils, fuels, chemicals, pharmaceuticals, paints and coatings.

Specifications

Quantity 500 mL
Type Color Reference Standard
View More Specs

Products 3
Product Code Brand Description Price Quantity Availability  
Product Code Brand Description Price Quantity Availability  
16020162
View Documents
Paragon Scientific
134200
ASTM Color 2 ASTM Color 2 N/A
N/A
16030162
View Documents
Paragon Scientific
134210
ASTM Color 5 ASTM Color 5 N/A
N/A
16040162
View Documents
Paragon Scientific
134220
ASTM Color 8 ASTM Color 8 N/A
N/A
Description

Description

  • Certified according to ISO 17025 / ISO 17034 under UKAS accreditation
  • Full Traceability to International Standards
  • 500mL Pack Size supplied with MSDS
  • 18 months shelf life
  • Tamper evident security packaging
  • Manufactured in the United kingdom
Specifications

Specifications

500 mL
Color Reference Standard
Videos
SDS
Documents

Documents

Product Certifications

Nominal values quoted