Learn More
Asecos™ G-Ultimate 90 Gas Cylinder Cabinet
Safe and approved cabinet for storage of flammable or toxic gases in working areas. Asecos™ G-Ultimate 90 Gas Cylinder Cabinet holds up to 3 x 50L gas cylinders.
Brand:  Asecos™ 30667-002-30668
Product Code. 13423877
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Robust and long-lasting
    • Triple-hinged door
    • Scratch- and impact-resistant
  • Easy handling
    • Doors stop in any position
    • Self-closing in the event of fire
  • Locking doors
  • Easy transport
  • Easy alignment
  • Ventilation
Specifications
Specifications
FM Approved, 30 EN 14470-1, EKS / AK4 / EN 14727, CE
616 mm
Storage of hazardous (flammable) substances in the workplace
1196 mm
Fire safety storage cabinet
3 Tray Shelves and Bottom Tray, 4 Full Extension Drawers, 6 Full Extension Drawers
616 x 1196 x 1968 mm
1968 mm
Light Gray
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Asecos™ G-Ultimate 90 Gas Cylinder Cabinet > Dimensions (D x W x H): 616 x 1196 x 1968mm