Learn More
Trajan Scientific and Medical Gas Filter Kit - FID
Prevent contaminants in gases from significantly impacting the quality of results. Oxygen, hydrocarbons and moisture can lead to problems such as noisy baselines, moisture entering the GC column, excessive bleed and septa degradation.
Brand: Trajan Scientific and Medical 1035164
Additional Details : Weight : 10.00000kg
Description
- Capacity of 150 mL oxygen, 7.2 g water, 7g organics depending upon impurities
- Indicator color change tells you when to change your filter
- Can be used in combination with other filters through a simple connecting unit
- Kit consists of gas filter and connecting unit
Specifications
150 mL | |
Carrier Gas | |
1 x 4-position connecting unit, 4 x gas filters (2 x hydrocarbon, 1 x oxygen, 1 x moisture) | |
1/4 in. |
For research use only