missing translation for 'onlineSavingsMsg'
Learn More

Gas Arc™ Gas Regulator

Encompass

POA - £99.24

Specifications

Type Gas Regulator
View More Specs

Products 3
Product Code Brand Gases Measured Price Quantity & Availability  
Product Code Brand Gases Measured Price Quantity & Availability  
12980840
Encompass
View Documents
Gas Arc™
VAA05002
POA
Each
View alternative products
Save Up to 
»
N/A
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.

Request a Price for this item here.

12970820
Encompass
View accessory products View Documents
Gas Arc™
FBA07052
Fuel Gas
POA
Each
View alternative products
Save Up to 
»
N/A
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.

Request a Price for this item here.

12900790
Encompass
View Documents
Gas Arc™
RAN01021
Carbon Dioxide N/A N/A N/A View alternative products
Save Up to 
»
Description

Description

  • Regulators to BS EN ISO 9001 and BS EN ISO 2503
  • Two gauge single and multi stage
  • Cylinder pressure up to 300bar
  • Robust brass construction to meet industry requirements
  • Tamper proof high flow safety relief valve
  • Safety gauges to BS EN ISO 562
  • Encapsulated valve design fitted with 316L sintered stainless steel filter
  • Regulator outlet connection: 3/8 inch BSP
  • Propane regulator is plugged
Specifications

Specifications

Gas Regulator
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
Gas Arc™ Gas Regulator

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.