Learn More
Lenz Laborglasintrumente Gas Washing Bottles Acc. to Drechsel with Filter Disc
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 05510237
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10214151
  • £84.00 / Each
Estimated Shipment: 19-08-2024
to see stock.
Add to basket
Description
Description
  • Acc. to DIN 12596, complete with head (with filter disc)
  • Ground joint connection: NS 29/32
  • Material: DURAN™ tubing (head), borosilicate glass 3.3 (bottle)
Specifications
Specifications
100 mL
Olive connections
NS 29/32
Borosilicate Glass (3.3), Duran Tubing
2