missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Cover for Precision™ Water Baths
Complement your water baths with a gassing hood, gable cover, designed to fit large flasks, or concentric ring cover for steaming.
Brand: Thermo Scientific™ GPSSL02
This item is not returnable.
View return policy
Description
Complement your water baths with a gassing hood, gable cover, designed to fit large flasks, or concentric ring cover for steaming.
Features:
Gassing Hoods
- Permit control over the atmosphere surrounding the sample
- Improve temperature uniformity and reduce energy consumption
- Polished stainless steel with plastic gas fitting
- Each bath will accommodate 1 large hood or 2 small hoods
Gable Covers
- Enhanced hinged design supports open-lid configuration and easy lid removal
- Accommodate taller labware with extended height
- Polycarbonate and stainless steel models available
- Warranty varies by country, please contact us for details
Services
1 year
Specifications
Stainless Steel Gable Cover | |
Precision 2L General Purpose Water Bath | |
Stainless Steel |
2 L | |
Gable Cover |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction