missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GATA-2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 6 publications
£238.00 - £424.00
Specifications
Antigen | GATA-2 |
---|---|
Concentration | 0.6mg/mL |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18482801
|
Novus Biologicals
NBP1-82581-25ul |
25 μL |
£238.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18791704
|
Novus Biologicals
NBP1-82581 |
0.1 mL |
£424.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GATA-2 Polyclonal specifically detects GATA-2 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
GATA-2 | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
Unconjugated | |
RUO | |
PBS (pH 7.2), 40% Glycerol with 0.02% Sodium Azide | |
endothelial transcription factor GATA-2, FLJ45948, GATA binding protein 2, GATA-binding protein 2NFE1B, MGC2306 | |
GATA2 | |
IgG | |
Affinity Purified | |
Specificity of human GATA-2 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
0.6mg/mL | |
Polyclonal | |
Rabbit | |
Human, Mouse | |
P23769 | |
2624 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:LTGGQMCRPHLLHSPGLPWLDGGKAALSAAAAHHHNPWTVSPFSKTPLHPSAAGGPGGPLSVYPGAGGGSGGGSGSSVASLTPTAAHSGSHLFGFPPTPPKEVSPDPSTTGAASPASSSAGGSAARG | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
51 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
GATA-2 Antibody, Novus Biologicals™