missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GATA-2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 6 publications
Brand: Novus Biologicals NBP1-82581-25ul
190.40 GBP valid until 2025-03-28
Use promo code "25306" to get your promotional price.
This item is not returnable.
View return policy
Description
GATA-2 Polyclonal specifically detects GATA-2 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
GATA-2 | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.2), 40% Glycerol with 0.02% Sodium Azide | |
endothelial transcription factor GATA-2, FLJ45948, GATA binding protein 2, GATA-binding protein 2NFE1B, MGC2306 | |
Rabbit | |
51 kDa | |
25 μL | |
Primary | |
Specificity of human GATA-2 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
0.6mg/mL | |
Western Blot 0.04-0.4 ug/mL, Simple Western 1:100, Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/Immunofluorescence 0.25 - 2 ug/mL, Immunoprecipitation, Immunohistochemistry-Paraffin 1:20 - 1:50, Immunohistochemistry-Frozen 1:200 - 1:500 | |
P23769 | |
GATA2 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:LTGGQMCRPHLLHSPGLPWLDGGKAALSAAAAHHHNPWTVSPFSKTPLHPSAAGGPGGPLSVYPGAGGGSGGGSGSSVASLTPTAAHSGSHLFGFPPTPPKEVSPDPSTTGAASPASSSAGGSAARG | |
Affinity Purified | |
RUO | |
2624 | |
Human, Mouse | |
IgG |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
GATA-2 Antibody, Novus Biologicals™ > 25 μL; Unlabeled
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction