Learn More
Thermo Scientific™ MAS™ Liquimmune™ Immunoassay Controls
Get relief from time consuming and costly QC standard lot changes for up to 5 years with ready-to-use, liquid assayed Thermo Scientific™ MAS™ Liquimmune™ Immunoassay Controls.
Brand: Thermo Scientific™ LIG-303
Description
Get relief from time-consuming and costly QC standard lot changes for up to 5 years with ready-to-use, liquid assayed Thermo Scientific™ MAS™ Liquimmune™ Immunoassay Controls. With a new 5-year shelf life, these multi-analyte controls simplify QC while monitoring immunoassay test procedures on most automated instruments. These controls are also available in a convenient Vista™ vial format. Extended claim may not apply for all countries.
- Features three significant levels
- Human serum matrix
- Assayed values available for most automated instruments

Specifications
3-year shelf life from date of manufacture at -25°C to -15°C; 90 day unopened-vial stability for all analytes at 2°C to 8°C; 30 day open-vial stability for all analytes at 2°C to 8°C. Extended 5-year shelf life beginning with lot LIA2004 in Fall 2015. | |
17-a-hydroxyprogesterone, Acetaminophen, alpha-Fetoprotein (AFP), Aldosterone, Amikacin, Benzodiazepines, Carbamazepine, Carcinoembryonic Antigen (CEA), Creatinine Kinase-MB (CK-MB), Cortisol, DHEA Sulfate, Digoxin, Disopyramide, Estradiol, Ethosuximide, Ferritin, Folate (Vitamin B9), Follicle Stimulating Hormone (FSH), Gentamicin, Growth Hormone, β-hCG, hCG, Homocysteine, IgA, IgE, IgM, LD-1, Luteinizing Hormone (LH), Lidocaine, Lithium, N-acetylprocainamide (NAPA), Phenobarbital, Phenytoin, Primidone, Procainamide, Progesterone, Prolactin, PSA, PSA (Free), Quinidine, Salicylate (SALY), T-uptake, T3, T3 (Free), T4, Testosterone, Theophylline, Thyroglobulin, Tobramycin, Tricyclic Antidepressants, Thyroid Stimulating Hormone (TSH), Valproic Acid, Vancomycin, Vitamin B12 | |
Control | |
6 Pk. |
Liquimmune Controls, Assayed | |
Serum | |
3 | |
5 mL |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
LIA28023 | Certificate of Analysis | 29/01/2025 | LIG-303 |
LIA24093 | Certificate of Analysis | 29/01/2020 | LIG-303 |
LIA22033 | Certificate of Analysis | 20/09/2019 | LIG-303 |
LIA20043 | Certificate of Analysis | 29/08/2019 | LIG-303 |
LIA16033-COA | Certificate of Analysis | 14/04/2017 | LIG-303 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.