Learn More
Thermo Scientific™ MAS™ Liquimmune™ Immunoassay Controls
Get relief from time consuming and costly QC standard lot changes for up to 5 years with ready-to-use, liquid assayed Thermo Scientific™ MAS™ Liquimmune™ Immunoassay Controls.
£183.00
Specifications
Sample Type Serum
Type Control
Packaging 6 Pk.
View More Specs
Products 3
Product Code Brand Description Level Price Quantity & Availability  
Product Code Brand Description Level Price Quantity & Availability  
17372733
View Documents
Thermo Scientific™
LIG-202
Liquimmune Controls, Assayed 2
£183.00
Each
17382733
View Documents
Thermo Scientific™
LIG-303
Liquimmune Controls, Assayed 3
£183.00
Each
17362733
View Documents
Thermo Scientific™
LIG-101
Liquimmune Controls, Assayed 1
£183.00
Each
Description
Description
Get relief from time-consuming and costly QC standard lot changes for up to 5 years with ready-to-use, liquid assayed Thermo Scientific™ MAS™ Liquimmune™ Immunoassay Controls. With a new 5-year shelf life, these multi-analyte controls simplify QC while monitoring immunoassay test procedures on most automated instruments. These controls are also available in a convenient Vista™ vial format. Extended claim may not apply for all countries.
  • Features three significant levels
  • Human serum matrix
  • Assayed values available for most automated instruments
  • Extended 5-year shelf life beginning with lot LIA2004 in Fall 2015
Specifications
Specifications
Serum
Control
6 Pk.
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Thermo Scientific™ MAS™ Liquimmune™ Immunoassay Controls