Learn More
Gatifloxacin sesquihydrate, Thermo Scientific Chemicals
A broad spectrum antibiotic
£18.69 - £221.00
Chemical Identifiers
CAS | 180200-66-2 |
---|---|
Molecular Formula | C19H22FN3O4 |
Molecular Weight (g/mol) | 375.40 |
MDL Number | MFCD00895399 |
InChI Key | XUBOMFCQGDBHNK-UHFFFAOYNA-N |
PubChem CID | 5379 |
ChEBI | CHEBI:5280 |
IUPAC Name | 1-cyclopropyl-6-fluoro-8-methoxy-7-(3-methylpiperazin-1-yl)-4-oxoquinoline-3-carboxylic acid |
SMILES | COC1=C2N(C=C(C(O)=O)C(=O)C2=CC(F)=C1N1CCNC(C)C1)C1CC1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15497229
|
Thermo Scientific Alfa Aesar
J66036.03 |
1 g |
£18.69
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15407239
|
Thermo Scientific Alfa Aesar
J66036.06 |
5 g |
£65.43
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15417239
|
Thermo Scientific Alfa Aesar
J66036.14 |
25 g | N/A | ||||||
Description
A broad spectrum antibiotic
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
180200-66-2 | |
375.40 | |
XUBOMFCQGDBHNK-UHFFFAOYNA-N | |
CHEBI:5280 | |
COC1=C2N(C=C(C(O)=O)C(=O)C2=CC(F)=C1N1CCNC(C)C1)C1CC1 |
C19H22FN3O4 | |
MFCD00895399 | |
5379 | |
1-cyclopropyl-6-fluoro-8-methoxy-7-(3-methylpiperazin-1-yl)-4-oxoquinoline-3-carboxylic acid |
Specifications
Gatifloxacin Sesquihydrate | |
180200-66-2 | |
C19H22FN3O4 | |
684°C | |
Soluble in 1N NaOH at 10mg/mL | |
COC1=C2N(C=C(C(O)=O)C(=O)C2=CC(F)=C1N1CCNC(C)C1)C1CC1 | |
375.40 | |
CHEBI:5280 | |
Gatifloxacin sesquihydrate |
Powder | |
White to Yellow | |
MFCD00895399 | |
14,4376 | |
XUBOMFCQGDBHNK-UHFFFAOYNA-N | |
1-cyclopropyl-6-fluoro-8-methoxy-7-(3-methylpiperazin-1-yl)-4-oxoquinoline-3-carboxylic acid | |
5379 | |
402.42 |
Safety and Handling
GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332
missing translation for 'rtecsNumber' : VB1983600
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
- Gatifloxacin sesquihydrate
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
- MIXTURE LIST-Contains : Gatifloxacin sesquihydrate
RUO – Research Use Only