missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Gay-Lussac Uncalibrated Density Bottles

Gay-Lussac density bottle made with Borosilicate Glass 3.3 that is ideal in most research, scientific, and other laboratory environments.

Brand:  BRAND™ 43220

68.54 GBP valid until 2024-12-31
Use promo code "21693" to get your promotional price.


 View more versions of this product

Product Code. 11818291

  • £85.85 / Pack of 2
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • BRAND™ density bottle
  • Gay-Lussac pattern
  • Uncalibrated capacity, 10 mL
  • Borosilicate glass, bottle narrow-mouth, bottle round (Gay-Lussac), white bottle
  • DIN ISO 3507
  • Stopper NS 10/19 with capillary
  • Top of stopper ground and polished
TRUSTED_SUSTAINABILITY
Specifications

Specifications

2Each
25 mL
Borosilicate Glass (3.3)
Gay-Lussac Density Bottle
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.