missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ Gay-Lussac Uncalibrated Density Bottles
Gay-Lussac density bottle made with Borosilicate Glass 3.3 that is ideal in most research, scientific, and other laboratory environments.
Brand: BRAND™ 43238
113.05 GBP valid until 2024-12-31
Use promo code "21693" to get your promotional price.
This item is not returnable.
View return policy
Description
- BRAND™ density bottle
- Gay-Lussac pattern
- Uncalibrated capacity, 10 mL
- Borosilicate glass, bottle narrow-mouth, bottle round (Gay-Lussac), white bottle
- DIN ISO 3507
- Stopper NS 10/19 with capillary
- Top of stopper ground and polished
Specifications
2Each | |
100 mL | |
Borosilicate Glass (3.3) | |
Gay-Lussac Density Bottle |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction