missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HiScreen™ Sepharose™ Prepacked High Performance HIC Columns
Designed for method optimization and parameter screening
Brand: Cytiva 28-9505-16
This item is not returnable.
View return policy
Description
- Ready-to-use columns for convenient process development optimization
- Standard aliphatic/aromatic Hydrophobic Interaction Chromatography media for capture and intermediate purification
- Bed height of 10cm designed to allow method optimization and parameter screening
- Easily connected in series to achieve 20cm bed height
- Small bed volume (4.7mL) gives fast result and minimal sample/buffer consumption
- Reproducible results, scalable to BioProcess columns packed with the same chromatography media using the same linear fluid velocity
HiScreen Phenyl HP
- Degree of substitution: ∽25μmol phenyl/mL gel
- Degree of substitution: ∽50μmol phenyl/mL gel
Specifications
1M NaOH, 1M Acetic Acid, 30% Aceto nitrile, 70% Ethanol, 30% Isopropanol, 6M Guanidine Hydrochloride, 8M Ure, 3M Ammoniumsulfate, Ionic Detergents, Non ionic Detergents, Polar Organic Solvents, 2% Sodium Dodectyl Sulfate | |
4.7mL | |
7.7mm | |
3.93 in. | |
<150cm/hr. | |
Phenyl | |
3 to 13 | |
6% Cross-linked Agarose | |
Ion Exchange Column |
HiScreen Phenyl Sepharose HP column | |
0.3 in. | |
1 x 4.7mL | |
100mm | |
Polypropylene | |
3 bar | |
24 to 44μm | |
4° to 30°C | |
Convenient process development optimization parameter screening |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- HiScreen Phenyl HP
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
- UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.