missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Gelatin Sepharose™ 4B Media

Product Code. 11544955 Shop All Cytiva Products
Click to view available options
Quantity:
25 mL
Unit Size:
25mL
This item is not returnable. View return policy

Product Code. 11544955

Brand: Cytiva 17095601

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Designed for the purification or depletion of fibronectin as gelatin binds to fibronectin with high specificit

  • Bovine gelatin coupled to Sepharose 4B, in a 4% agarose matrix
  • Ligand coupling method: cyanogen bromide activation
  • Ligand concentration: 4.5-8mg gelatin/mL medium
  • Average particle size: 90μm
  • Stable in the pH range 3 to 10, and in the presence of all commonly used buffers

TRUSTED_SUSTAINABILITY

Specifications

Quantity 25 mL
Material Gelatin Sepharose 4 B
Particle Size 90 μm
Matrix Affinity Resin
compliance-icons
Product Identifier
  • gelatin sepharose 4B
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : ethanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.