Learn More
Thermo Scientific™ GeneBLAzer™ ADRA1A CHO-K1 DA Assay Kit
The GeneBLAzer™ ADRA1A CHO-K1 DA (Division Arrested) cells contain the human Adrenergic Alpha-1A Receptor (ADRA1A), stably integrated into the CellSensor™ NFAT-bla CHO-K1 cell line.
Brand:  Thermo Scientific™ K1577
Product Code. 10465575
  • £608.00 / Each
Estimated Shipment: 16-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
The GeneBLAzer™ ADRA1A CHO-K1 DA (Division Arrested) cells contain the human Adrenergic Alpha-1A Receptor (ADRA1A), (Accession No. NM_000680) stably integrated into the CellSensor™ NFAT-bla CHO-K1 cell line. CellSensor™ NFAT-bla CHO-K1 cells (Cat. No. K1078) contain a beta-lactamase (bla) reporter gene under control of the NFAT response element.
Both ADRA1A CHO-K1 DA cells are functionally validated for Z'-factor and EC50 concentrations of Phenylephrine. In addition, ADRA1A-NFAT-bla CHO-K1 cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary in the Manuals and Brochures section. DA cells are irreversibly division arrested using a low-dose treatment of Mitomycin C, and have no apparent toxicity or change in cellular signal transduction.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NM_000680
Division-Arrested Cells
GeneBLAzer™ ADRA1A CHO-K1 DA Assay Kit, 1 Plate, ∽4,800,000 cells/vial Quantity of Cells, CHO-K1 Cell Line, Fluorescent Detection Method, GPCRs Druggable Target, BLA (Beta-Lactamase) Reporter, GeneBLAzer™ System, Liquid Nitrogen Storage
Pharma and biopharma, drug discovery and development, target and lead identification and validation, G-protein coupled receptor biology, cell-based GPCR reporter assays
Each kit contains sufficient quantities of division arrested (DA) cells and substrate to assay 1 x 384 well plate. (Other materials are required separately; please refer to the protocol). Includes: GeneBLAzer™ ADRA1A CHO-K1DA cells, LiveBLAzer™-FRET B/G Loading Kit, 70μg, Solution D, GeneBLAzer™ ADRA1A CHO-K1Assay Protocol, LiveBLAzer™-FRET B/G Loading Protocol, Certificate of Analysis
GeneBLAzer™
BLA (Beta-Lactamase)
ADRA1A
CHO-K1
GPCR
Cell-based beta lactamase reporter gene
Each kit contains sufficient quantities of division arrested (DA) cells and substrate to assay 1 x 384 well plate. (Other materials are required separately; please refer to the protocol).
Includes:
• GeneBLAzer™ ADRA1A CHO-K1DA cells
• LiveBLAzer™-FRET B/G Loading Kit, 70 μg
• Solution D
• GeneBLAzer™ ADRA1A CHO-K1Assay Protocol
• LiveBLAzer™-FRET B/G Loading Protocol
• Certificate of Analysis
The GeneBLAzer™ ADRA1A CHO-K1 DA cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.
Fluorescence
ADRA1A
Pool of Stable Cells
1 Plate
CHO-K1
Division-Arrested Cells Assay Kit
GeneBLAzer™