Learn More
Thermo Scientific™ GeneBLAzer™ ADRA1A-NFAT-bla CHO-K1 Cells
The GeneBLAzer™ ADRA1A-NFAT-bla CHO-K1 cells contain the human Adrenergic Alpha-1A Receptor (ADRA1A), (Accession No. NM_000680) stably integrated into the CellSensor™ NFAT-bla CHO-K1 cell line.
Brand: Thermo Scientific™ K1470
Description
The GeneBLAzer™ ADRA1A-NFAT-bla CHO-K1 cells contain the human Adrenergic Alpha-1A Receptor (ADRA1A), (Accession No. NM_000680) stably integrated into the CellSensor™ NFAT-bla CHO-K1 cell line. CellSensor™ NFAT-bla CHO-K1 cells (Cat No. K1078) contain a beta-lactamase (bla) reporter gene under control of the NFAT response element.
Both ADRA1A-NFAT-bla CHO-K1 cells are functionally validated for Z'-factor and EC50 concentrations of Phenylephrine. In addition, ADRA1A-NFAT-bla CHO-K1 cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary in the Manuals and Brochures section.
Specifications
NM_000680 | |
Dividing Cells | |
Fluorescence | |
ADRA1A | |
Pool of Stable Cells | |
2 x 106 cells | |
ADRA1A | |
CHO-K1 | |
GPCR |
Cell-based beta lactamase reporter gene | |
LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol. Includes: • GeneBLAzer™ ADRA1A -NFAT-bla CHO-K1 cells • GeneBLAzer™ ADRA1A -NFAT-bla CHO-K1 Assay Protocol • Certificate of Analysis The GeneBLAzer™ ADRA1A -NFAT-bla CHO-K1 cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt. |
|
GPCRs | |
ADRA1A -NFAT-bla CHO-K1 cells, ADRA1A -NFAT-bla CHO-K1 Assay Protocol, Certificate of Analysis | |
GeneBLAzer™ | |
BLA (Beta-Lactamase) | |
CHO-K1 Cells | |
GeneBLAzer™ |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
For Research Use Only. Not for use in diagnostic procedures.