Learn More
Thermo Scientific™ GeneBLAzer™ ADRB2-CRE-bla CHO-K1 Cells
The GeneBLAzer™ ADRB2-CRE-bla CHO-K1 cells contain the human Adrenergic Beta-2 Receptor (ADRB2), (Accession No. NM_000024.3) stably integrated into the CellSensor™ CRE-bla CHO-K1 cell line.
Brand: Thermo Scientific™ K1472
Description
The GeneBLAzer™ ADRB2-CRE-bla CHO-K1 cells contain the human Adrenergic Beta-2 Receptor (ADRB2), (Accession No. NM_000024.3) stably integrated into the CellSensor™ CRE-bla CHO-K1 cell line. CellSensor™ CRE-bla CHO-K1 cells (Cat. No. K1129) contain a beta-lactamase (bla) reporter gene under control of the cAMP response element (CRE).
The ADRB2-CRE-bla CHO-K1 cells are functionally validated for Z'-factor and EC50 concentrations of Isoproterenol. In addition, ADRB2-CRE-bla CHO-K1 cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary in the Manuals and Brochures section.
Specifications
NM_000024.3 | |
Dividing Cells | |
Fluorescence | |
ADRB2 | |
Single Cell Clone Origin | |
2 x 106 Cells | |
ADRB2 | |
CHO-K1 | |
GPCR |
Cell-based beta lactamase reporter gene | |
LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol. Includes: • GeneBLAzer™ ADRB2-CRE-bla CHO-K1 cells • GeneBLAzer™ ADRB2-CRE-bla CHO-K1 Assay Protocol • Certificate of Analysis The GeneBLAzer™ ADRB2-CRE-bla CHO-K1 cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt. |
|
GPCRs | |
ADRB2-CRE-bla CHO-K1 cells, ADRB2-CRE-bla CHO-K1 Assay Protocol, Certificate of Analysis | |
GeneBLAzer™ | |
BLA (Beta-Lactamase) | |
CHO-K1 Cells | |
GeneBLAzer™ |
For Research Use Only. Not for use in diagnostic procedures.