missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneBLAzer™ AVPR1A CHO-K1 DA Assay Kit

GeneBLAzer™ AVPR1A CHO-K1 DA (Division Arrested) cells and AVPR1A-NFAT-bla CHO-K1 cells contain the human Arginine Vasopressin 1a (AVPR1A) receptor stably integrated into the CellSensor™ NFAT-bla CHO-K1 cell line.

Brand:  Thermo Scientific™ K1323

Product Code. 10454035

  • £618.00 / Each
Estimated Shipment: 16-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

This cell line is available as dividing cells by requesting a quote for Cat. No. K2186 from dividingcells@invitrogen.com

GeneBLAzer™ AVPR1A CHO-K1 DA (Division Arrested) cells and AVPR1A-NFAT-bla CHO-K1 cells contain the human Arginine Vasopressin 1a (AVPR1A) receptor stably integrated into the CellSensor™ NFAT-bla CHO-K1 cell line. CellSensor™ NFAT-bla CHO-K1 cells (Cat. no.K1129) contain a beta-lactamase reporter gene under control of the nuclear factor of activated T-cells (NFAT) response element. Division Arrested (DA) cells are available in two configurations- an Assay Kit (which includes cells and sufficient substrate to analyze 1 x 384-well plate), and a tube of cells sufficient to analyze 10 x 384-well plates.

DA cells are irreversibly division arrested using a low-dose treatment of Mitomycin-C, and have no apparent toxicity or change in cellular signal transduction. Both AVPR1A CHO-K1 DA cells and AVPR1A-NFAT-bla CHO-K1 cells are functionally validated for Z'-factor and EC50 concentrations of vasopressin, (Figure 1). In addition, AVPR1A-NFAT-bla CHO-K1 cells have been tested for assay performance under variable conditions, including DMSO concentration, cell number, stimulation time, and substrate loading time (data available upon request). Additional testing data using alternate stimuli are also available.

The family of receptors that recognize vasopressin and oxytocin consists of four members: vasopressin receptors 1a (AVPR1a), 1b (AVPR1b), and 2 (AVPR2), and the oxytocin (OT) receptor. AVPR1a, AVPR1b, and the OT receptor couple to the Gq/G11 class of G proteins and trigger the release of intracellular calcium when stimulated. AVPR2 receptors couple to Gs proteins and activate adenyl cyclase. All three AVPR receptors have high affinity for their endogenous ligand, vasopressin, but AVPR1b has less affinity for oxytocin than does AVPR1a or AVPR2. The hormones vasopressin and oxytocin are synthesized in the hypothalamus. Oxytocin causes contractions in the uterus during childbirth. The main functions of arginine vasopressin (AVP) are reabsorption of water by the kidneys and contraction of smooth muscle in arteries. This system may also play a role in memory and learning.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

NM_000706.3
Division-Arrested Cells
Gene ID: 552
GPCRs
AVPR1a
GeneBLAzer™
BLA (Beta-Lactamase)
Division-Arrested Cells Assay Kit
GeneBLAzer™
Cell-based beta lactamase reporter gene
Liquid Nitrogen
Fluorescence
552
Single Cell Clone Origin
1 Plate
AVPR1A
CHO-K1
GPCR
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.