Learn More
Thermo Scientific™ GeneBLAzer™ Bradykinin-B2-NFAT-bla CHO-K1 Cells
GeneBLAzer™ Bradykinin-B2-NFAT-bla CHO-K1 cells contain the human Bradykinin Receptor B2 (B2) stably integrated into the CellSensor™ CRE-bla CHO-K1 cell line.
Brand: Thermo Scientific™ K1790
Description
GeneBLAzer™ Bradykinin-B2-NFAT-bla CHO-K1 cells contain the human Bradykinin Receptor B2 (B2) stably integrated into the CellSensor™ CRE-bla CHO-K1 cell line. CellSensor™ NFAT-bla CHO-K1 cells (Cat. No. K1534) contain a beta-lactamase reporter gene under control of the Cyclic AMP Response Element (CRE) response element.
The Bradykinin-B2-NFAT-bla CHO-K1 cells are functionally validated for Z' and EC50 concentrations of Bradykinin. In addition, GeneBLAzer™ Bradykinin-B2-NFAT-bla CHO-K1 cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary.
For Academic pricing please login or contact us at discoverysciences@invitrogen.com.
Specifications
NM_000623.2 | |
Dividing Cells | |
GeneBLAzer™ Bradykinin-B2-NFAT-bla CHO-K1 Cells, 8 x 106 Cells, CHO-K1 Cell Line, Fluorescent Detection Method, GPCRs Druggable Target, BLA (Beta-Lactamase) Reporter, FRET Technique, Liquid Nitrogen Storage | |
FRET | |
K1790 GeneBLAzer™ Bradykinin-B2-NFAT-bla CHO-K1 cells (LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol). Includes: GeneBLAzer™ Bradykinin-B2-NFAT-bla CHO-K1 cells, GeneBLAzer™ Bradykinin-B2-NFAT-bla CHO-K1 Assay Protocol, Certificate of Analysis | |
GeneBLAzer™ | |
End Point | |
CHO-K1 | |
CHO-K1 Cells | |
GeneBLAzer™ |
Cell-based beta lactamase reporter gene | |
LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol. Includes: • GeneBLAzer™ Bradykinin-B2-NFAT-bla CHO-K1 cells • GeneBLAzer™ Bradykinin-B2-NFAT-bla CHO-K1 Assay Protocol • Certificate of Analysis The GeneBLAzer™ Bradykinin-B2-NFAT-bla CHO-K1 cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt. |
|
Fluorescence | |
BK2 | |
Single Cell Clone Origin | |
8 x 106 Cells | |
BLA (Beta-Lactamase) | |
BDKRB2 | |
CHO-K1 | |
GPCR |
For Research Use Only. Not for use in diagnostic procedures.