Learn More
Thermo Scientific™ GeneBLAzer™ EDG3-Gα15 HEK 293T DA Assay Kit
GeneBLAzer™ EDG3-Gα15 HEK 293T DA (Division Arrested) cells and EDG3-Gα15-NFAT-bla HEK 293T cells contain the human EDG3 receptor stably integrated in the GeneBLAzer™ Gα15-NFAT-bla HEK 293T cell line.
Brand: Thermo Scientific™ K1319
Description
This cell line is available as dividing cells by requesting a quote for K1234 from dividingcells@invitrogen.com
GeneBLAzer™ EDG3-Gα15 HEK 293T DA (Division Arrested) cells and EDG3-Gα15-NFAT-bla HEK 293T cells contain the human EDG3 receptor stably integrated in the GeneBLAzer™ Gα15-NFAT-bla HEK 293T cell line. GeneBLAzer™ Gα15-NFAT-bla HEK 293T cells (Cat. No. K1212) contain a beta-lactamase reporter gene under control of the Nuclear Factor of Activated T-cells (NFAT) response element, and stably express the promiscuous G-protein, Gα15. Division Arrested (DA) cells are available in two configurations: an Assay Kit (which includes cells and sufficient substrate to analyze 1 x 384-well plate), and a tube of cells sufficient to analyze 10 x 384-well plates.
DA cells are irreversibly division arrested using a low-dose treatment of Mitomycin-C, and have no apparent toxicity or change in cellular signal transduction. Both EDG3-Gα15 HEK 293T DA cells and EDG3-Gα15-NFAT-bla HEK 293T cells are functionally validated for Z'-factor and EC50 concentrations of Sphingosine-1-phosphate (S1P), (Figure 1). In addition, EDG3-Gα15-NFAT-bla HEK 293T cells have been tested for assay performance under variable conditions, including DMSO concentration, cell number, stimulation time, and substrate loading time (data available upon request). Additional testing data using alternate stimuli are also available.
EDG-3(Endothelial-differentiation-gene-3)/S1P-3(Sphingosine-1-Phosphate-3) is a Gq/Gi/Go/G13/G12 coupled GPCR. EDG-3 has been shown to be responsible for bradycardia that has been induced in clinical trials as a side effect of the immunosuppressive drug FTY720 that targets EDG-1. EDG-3 has also been shown to induce vasodilation in response to sphingosylphosphorylcholine (SPC), sphingosine-1-phosphate (S1P), and lysosulfatide (LSF). The EDG-3 Receptor has also been shown to play a cooperative or redundant role in angiogenesis with EDG-1
Specifications
NM_005226.2 | |
Division-Arrested Cells | |
Gene ID: 1903 | |
GPCRs | |
EDG3 | |
GeneBLAzer™ | |
BLA (Beta-Lactamase) | |
Division-Arrested Cells Assay Kit | |
GeneBLAzer™ |
Cell-based beta lactamase reporter gene | |
GeneBLAzer™ EDG3-Gα15 HEK 293T DA Assay Kit Each system contains sufficient division arrested (DA) cells & substrate to assay 1 x 384-well plate. (Other materials are required separately; please refer to the protocol). • EDG3-Gα15 HEK 293T DA cells (K1319A) • LiveBLAzer™-FRET B/G Loading Kit, 70 μg • Solution D, 1 mL • 2 protocols • Certificate of Analysis GeneBLAzer™ EDG3-Gα15 HEK 293T DA cells Each system contains sufficient division arrested (DA) cells to assay 10 x 384-well plates (LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol). • EDG3-Gα15 HEK 293T DA cells (K1320A) • 1 protocol • Certificate of Analysis GeneBLAzer™ EDG3-Gα15-NFAT-bla HEK 293T cells (LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol). • EDG3-Gα15-NFAT-bla HEK 293T cells (K1234) • 1 protocol • Certificate of Analysis Storage: GeneBLAzer™ EDG3-Gα15 HEK 293T DA Cells and EDG3-Gα15-NFAT-bla HEK 293T Cells are shipped on dry ice. Store in liquid nitrogen immediately upon receipt or thaw for immediate use. |
|
Fluorescence | |
1903 | |
Single Cell Clone Origin | |
1 plate | |
EDG3 | |
293T (HEK) | |
GPCR |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.