missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneBLAzer™ EDNRA-NFAT-bla HEK 293T Cells

GeneBLAzer™ EDNRA-NFAT-bla HEK293T cells contain the human Endothelin Type A Recpetor (EDNRA) stably integrated into the CellSensor™ NFAT-bla HEK293T cell line.

Brand:  Thermo Scientific™ K1726

Product Code. 10591814

  • £11140.00 / Each
Estimated Shipment: 20-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

CellSensor™ NFAT-bla HEK293T cells (Cat. no. K1538) contain a beta-lactamase reporter gene under control of the Nuclear Factor of Activated T-cells (NFAT) response element.

The GeneBLAzer™ EDNRA-NFAT-bla HEK293T cells are functionally validated for Z' and EC50 concentrations of ET-1. In addition, GeneBLAzer™ EDNRA-NFAT-bla HEK293T cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary.

The ENDRA receptor mediates signaling of endothelin proteins that regulate several critical biological processes, including development and function of blood vessels, production of certain hormones, and stimulation of cell growth and division. Defects in EDNRA-mediated signaling have been implicated in a wide range of diseases, including cancer, hypertension, and cardiovascular disorders.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

NM_001957
Dividing Cells
Fluorescence
EDNRA
GeneBLAzer™
BLA (Beta-Lactamase)
293T (HEK)
GPCR
Cell-based beta lactamase reporter gene
Contents:
K1726 GeneBLAzer™ EDNRA-NFAT-bla HEK293T cells
(LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol).
Includes:
• GeneBLAzer™ EDNRA-NFAT-bla HEK293T cells
• GeneBLAzer™ EDNRA-NFAT-bla HEK293T Assay Protocol
• Certificate of Analysis

Storage:
The GeneBLAzer™ EDNRA-NFAT-bla HEK293T cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.
FRET
Single Cell Clone Origin
2 x 106 Cells
EDNRA
GeneBLAzer™
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.