missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneBLAzer™ FPRL-1-Gα15-NFAT-bla CHO-K1 Cells

Product Code. 10217133
Click to view available options
Quantity:
2 x 106 Cells
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10217133

Brand: Thermo Scientific™ K1532

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

GeneBLAzer™ FPRL-1-Gα15-NFAT-bla CHO-K1 cells contain the human Formyl Peptide Like Receptor 1 (FPRL-1) stably integrated into the GeneBLAzer™ Gα15-NFAT-bla CHO-K1 cell line.

GeneBLAzer™ FPRL-1-Gα15-NFAT-bla CHO-K1 cells contain the human Formyl Peptide Like Receptor 1 (FPRL-1) stably integrated into the GeneBLAzer™ Gα15-NFAT-bla CHO-K1 cell line. GeneBLAzer™ Gα15-NFAT-bla CHO-K1 cells (Cat. No. K1537) contain a beta-lactamase reporter gene under control of the Nuclear Factor of Activated T-cells (NFAT) response element and the Promiscuous G protein, Gα15.

The GeneBLAzer™ FPRL-1-Gα15-NFAT-bla CHO-K1 cells are functionally validated for Z' and EC50 concentrations of W Peptide. In addition, GeneBLAzer™ FPRL-1-Gα15-NFAT-bla CHO-K1 cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary.

The FPRL-1 receptor has been implicated in a variety of inflammation and neurological disorders, including Alzheimer's disease.

For Academic pricing please login or contact us at discoverysciences@invitrogen.com.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number NM_001462
Assay Entry Cell-based beta lactamase reporter gene
Cell State Dividing Cells
Content And Storage LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol.

Includes:
• GeneBLAzer™ FPRL-1-Gα15-NFAT-bla CHO-K1 cells
• GeneBLAzer™ FPRL-1-Gα15-NFAT-bla CHO-K1 Assay Protocol
• Certificate of Analysis

The GeneBLAzer™ FPRL-1-Gα15-NFAT-bla CHO-K1 cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.
Detection Method Fluorescence
For Use With (Application) FRET
Gene Symbol FPRL-1
Includes FPRL-1-Gα15-NFAT-bla CHO-K1 cells, FPRL-1-Gα15-NFAT-bla CHO-K1 Assay Protocol, Certificate of Analysis
Population Status Single Cell Clone Origin
Product Line GeneBLAzer
Quantity 2 x 106 Cells
Reporter Gene BLA (Beta-Lactamase)
Target Entry FPRL-1
Type Dividing Cells
Cell Line CHO-K1
System Type GeneBLAzer™
Druggable Target GPCR
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.