Learn More
Invitrogen™ GeneBLAzer™ GPR10-NFAT-bla CHO-K1 Cells
GeneBLAzer™ GPR10-NFAT-bla CHO-K1 Cells
Brand: Invitrogen™ K1732
Code : NEW
Additional Details : Weight : 0.01000kg
Description
GeneBLAzer™ GPR10 CRE-bla CHO-K1 cells contain the human Prolactin Releasing Peptide (GPR10) stably integrated into the CellSensor™ CRE-bla CHO-K1 cell line. CellSensor™ CRE-bla CHO-K1 cells (Cat. no. K1535) contain a beta-lactamase reporter gene under control of the Cyclic AMP Response Element (CRE) response element.The GeneBLAzer™ GPR10 CRE-bla CHO-K1 cells are functionally validated for Z' and EC50 concentrations of PrRP20. In addition, GeneBLAzer™ GPR10 CRE-bla CHO-K1 cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary.
The GPR10 receptor has been implicated in a variety of metabolic disorders, including lactation, the regulation of food intake, and other physiological functions.
For Academic pricing please login or contact us at discoverysciences@invitrogen.com.
Specifications
-80°C | |
Pharma and biopharma, target and lead identification and validation | |
2 x 106 Cells | |
CHO-K1 Cells |
Fluorescent | |
GeneBLAzer™ GPR10 CRE-bla CHO-K1 cells, GeneBLAzer™ GPR10 CRE-bla CHO-K1 assay protocol, certificate of analysis | |
CHO-K1 |
For Research Use Only. Not for use in diagnostic procedures.