Learn More
Thermo Scientific™ GeneBLAzer™ GRPR-NFAT-bla CHO-K1 Cells
The GeneBLAzer™ GRPR-NFAT-bla CHO-K1 cells contain the human Gastrin Releasing Peptide Receptor (GRPR) stably integrated into the GeneBLAzer™ NFAT-bla CHO-K1 cell line.
Brand: Thermo Scientific™ K1561
Description
The GeneBLAzer™ GRPR-NFAT-bla CHO-K1 cells contain the human Gastrin Releasing Peptide Receptor (GRPR) stably integrated into the GeneBLAzer™ NFAT-bla CHO-K1 cell line. GeneBLAzer™ NFAT-bla CHO-K1 (Cat. No. K1447) contains a beta-lactamase (bla) reporter gene under control of a NFAT response element stably integrated into CHO-K1 cells.
The GRPR-NFAT-bla CHO-K1 cells are functionally validated for Z'-factor and EC50 concentrations using established ligands. In addition, GRPR-NFAT-bla CHO-K1 cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary.
Specifications
NM_005314.2 | |
Dividing Cells | |
Fluorescence | |
GRPR | |
Single Cell Clone Origin | |
2 x 106 Cells | |
GRPR | |
CHO-K1 | |
GPCR |
Cell-based beta lactamase reporter gene | |
LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol. Includes: • GeneBLAzer™ GRPR-NFAT-bla CHO-K1 cells • GeneBLAzer™ GRPR-NFAT-bla CHO-K1 Assay Protocol • Certificate of Analysis The GeneBLAzer™ GRPR-NFAT-bla CHO-K1 cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt. |
|
GPCRs | |
GRPR-NFAT-bla CHO-K1 cells, GRPR-NFAT-bla CHO-K1 Assay Protocol, Certificate of Analysis | |
GeneBLAzer™ | |
BLA (Beta-Lactamase) | |
Dividing Cells | |
GeneBLAzer™ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.