missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneBLAzer™ H1-NFAT-bla HEK 293T Cells

Product Code. 10405195
Click to view available options
Quantity:
2 x 106 Cells
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10405195

Brand: Thermo Scientific™ K1703

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

GeneBLAzer™ H1-NFAT-bla HEK293T cells contain the human Histamine Receptor 1 (H1) stably integrated into the CellSensor™ NFAT-bla HEK293T cell line.

GeneBLAzer™ H1-NFAT-bla HEK293T cells contain the human Histamine Receptor 1 (H1) stably integrated into the CellSensor™ NFAT-bla HEK293T cell line. CellSensor™ NFAT-bla HEK293T cells (Cat. No. K1538) contain a beta-lactamase reporter gene under control of the Cyclic AMP Response Element (CRE) response element.

The GeneBLAzer™ H1-NFAT-bla HEK293T cells are functionally validated for Z' and EC50 concentrations Histamine. In addition, GeneBLAzer™ H1-NFAT-bla HEK293T cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary.

The H1 receptor is located in human bronchi, bronchioles, and bronchial muscle, smooth muscle of the ileum, and the uterus. H1 receptors play a role in various disorders, including allergic reactions and mood disorders.

For Academic pricing please login or contact us at discoverysciences@invitrogen.com.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number NM_000861.3
Assay Entry Cell-based beta lactamase reporter gene
Cell State Dividing Cells
Content And Storage LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol.

Includes:
• GeneBLAzer™ H1-NFAT-bla HEK293T cells
• GeneBLAzer™ H1-NFAT-bla HEK293T Assay Protocol
• Certificate of Analysis

The GeneBLAzer™ H1-NFAT-bla HEK293T cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.
Detection Method Fluorescence
For Use With (Application) FRET
Gene Symbol H1
Includes H1-NFAT-bla HEK293T cells, H1-NFAT-bla HEK293T Assay Protocol, Certificate of Analysis
Population Status Single Cell Clone Origin
Product Line GeneBLAzer
Quantity 2 x 106 Cells
Reporter Gene BLA (Beta-Lactamase)
Target Entry HRH1, H1
Type Cell Line
Cell Line 293T (HEK)
System Type GeneBLAzer™
Druggable Target GPCR
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.