Learn More
Thermo Scientific™ GeneBLAzer™ M3-NFAT-bla CHO-K1 Cells
Description
GeneBLAzer™ M3-NFAT-bla CHO-K1 cells contain the human Acetylcholine (Muscarinic) Receptor 3 (M3) stably integrated into the CellSensor™ NFAT-bla CHO-K1 cell line. CellSensor™ NFAT-bla CHO-K1 cells (Cat. No. K1534) contain a beta-lactamase reporter gene under control of the Nuclear Factor of Activated T-cells (NFAT) response element.
The GeneBLAzer™ M3-NFAT-bla CHO-K1 cells are functionally validated for Z' and EC50 concentrations of Carbachol. In addition, GeneBLAzer™ M3-NFAT-bla CHO-K1 cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary.
The M3 receptor is localized in smooth muscle, exocrine glands, and the central nervous system. M3 has been implicated in autoimmune and neurological disorders.
For Academic pricing please login or contact us at discoverysciences@invitrogen.com.
Specifications
Specifications
| Accession Number | NM_000740.2 |
| Assay Entry | Cell-based beta lactamase reporter gene |
| Cell State | Dividing Cells |
| Content And Storage | LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol. Includes: • GeneBLAzer™ M3-NFAT-bla CHO-K1 cells • GeneBLAzer™ M3-NFAT-bla CHO-K1 Assay Protocol • Certificate of Analysis The GeneBLAzer™ M3-NFAT-bla CHO-K1 cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt. |
| Detection Method | Fluorescence |
| For Use With (Application) | FRET |
| Gene Symbol | M3 |
| Includes | M3-NFAT-bla CHO-K1 cells, M3-NFAT-bla CHO-K1 Assay Protocol, Certificate of Analysis |
| Population Status | Single Cell Clone Origin |
| Product Line | GeneBLAzer |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.