missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GeneBLAzer™ MC2R-CRE-bla-CHO-K1 Cells

Product Code. 10702154
Click to view available options
Quantity:
2 x 106 Cells
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10702154

Brand: Invitrogen™ K1483

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

GeneBLAzer™ MC2R-CRE-bla-CHO-K1 Cells

The GeneBLAzer™ MC2R-CRE-bla-CHO-K1 cells contain the human melanocortin-2 receptor (MC2R), (Accession # NM_000529.1) and the Melanocortin-2 receptor accessory protein (MRAP), (Accession #NM_178817.3) stably integrated into the CellSensor™ CRE-bla CHO-K1 cell line. CellSensor™ CRE-bla CHO-K1 cells (Cat # K1129) contain a beta-lactamase (bla) reporter gene under control of the cAMP Response Element (CRE).

The MC2R-CRE-bla-CHOK1 cells are functionally validated for Z'-factor and EC50 concentrations of ACTH 1-24. In addition, MC2R-CRE-bla-CHO-K1 cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary in the Manuals and Brochures section.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NM_000529.2
Assay Entry Cell-based beta lactamase reporter gene
Cell State Dividing Cells
Content And Storage (LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol).
Includes:
o GeneBLAzer ™ MC2R-CRE-bla-CHO-K1 cells
o GeneBLAzer ™ MC2R-CRE -bla CHO-K1 Assay Protocol
o Certificate of Analysis
The GeneBLAzer™ MC2R-CRE -bla CHO-K1 cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.
Detection Method Fluorescence
Gene Symbol MC2R
Population Status Single Cell Clone Origin
Product Line GeneBLAzer
Quantity 2 x 106 Cells
Reporter Gene BLA (Beta-Lactamase)
Target Entry MC2R
Type CHO-K1 Cells
Cell Line CHO-K1
System Type GeneBLAzer™
Druggable Target GPCR
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.