Learn More
Thermo Scientific™ GeneBLAzer™ NPSR1-B-NFAT-bla CHO-K1 Cells
The GeneBLAzer™ NPSR1-B-NFAT-bla CHO-K1 cells contain the human Neuropeptide S Receptor 1-Isoform B (NPSR1-B), (Accession # NP_997056) stably integrated into the CellSensor™ NFAT-bla CHO-K1 cell line.
Brand: Thermo Scientific™ K1481B
Description
CellSensor™ NFAT-bla CHO-K1 cells (Cat #K1078) contain a beta-lactamase (bla) reporter gene under control of the NFAT response element.
The NPSR1-B-NFAT-bla CHO-K1 cells are functionally validated for Z'-factor and EC50 concentrations of Neuropeptide-S. In addition, NPSR1-B-NFAT-bla CHO-K1 cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary in the Manuals and Brochures section.
Specifications
NP_997056.1 | |
(LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol). Includes: o GeneBLAzer™ NPSR1-B-NFAT-bla CHO-K1 cells o GeneBLAzer™ NPSR1-B-NFAT-bla CHO-K1 Assay Protocol o Certificate of Analysis The GeneBLAzer™ NPSR1-B-NFAT-bla CHO-K1 cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt. |
|
NPSR1-B | |
BLA (Beta-Lactamase) | |
CHO-K1 | |
GPCR |
Dividing Cells | |
Fluorescence | |
2 x 106 cells | |
CHO-K1 Cells | |
GeneBLAzer™ |