missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneBLAzer™ PAC1-CRE-bla CHO-K1 Cells

The GeneBLAzer™ PAC1-CRE-bla CHO-K1 cells contain the human Adenylate Cyclase Activating Polypeptide 1 Receptor 1 (ADCYAP1R/PAC1) stably integrated into the CellSensor™ CRE-bla CHO-K1 cell line.

Brand:  Thermo Scientific™ K1525

Product Code. 10093433

  • £10780.00 / Each
Estimated Shipment: 06-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

The PAC1-CRE-bla CHO-K1 cells are functionally validated for Z'-factor and EC50 concentrations of established ligands. In addition, PAC1-CRE-bla CHO-K1 cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary

TRUSTED_SUSTAINABILITY
Specifications

Specifications

NM_001118
Dividing Cells
Fluorescence
Single Cell Clone Origin
2 x 106 cells
ADCYAP1R1, PAC1
GeneBLAzer™
Cell-based beta lactamase reporter gene
K1525 GeneBLAzer™ PAC1-CRE-bla CHO-K1 cells
(LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol).
Includes:
• GeneBLAzer™ PAC1-CRE-bla CHO-K1 cells
• GeneBLAzer™ PAC1-CRE-bla CHO-K1 Assay Protocol
• Certificate of Analysis

Storage:
The GeneBLAzer™ PAC1-CRE-bla CHO-K1 cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.
ADCYAP1R1
GeneBLAzer™
BLA (Beta-Lactamase)
CHO-K1
GPCR
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.