missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneBLAzer™ PPARγ-UAS-bla HEK 293H Cells

PPAR gamma-UAS-bla HEK 293H cells contain the ligand-binding domain (LBD) of the human Peroxisome Proliferator-Activated Receptor-gamma (PPAR gamma) fused to the DNA-binding domain of GAL4 stably integrated in the GeneBLAzer™UAS-bla HEK 293H cell line.

Brand:  Thermo Scientific™ K1701

Product Code. 10175173

  • £7080.00 / Each
Estimated Shipment: 16-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

PPAR gamma-UAS-bla HEK 293H cells contain the ligand-binding domain (LBD) of the human Peroxisome Proliferator-Activated Receptor-gamma (PPAR gamma) fused to the DNA-binding domain of GAL4 stably integrated in the GeneBLAzer™UAS-bla HEK 293H cell line. GeneBLAzer™UAS-bla HEK 293H cells stably express a beta-lactamase reporter gene under the transcriptional control of an upstream activator sequence (UAS). When an agonist binds to the LBD of the GAL4 (DBD)-PPAR gamma (LBD) fusion protein, the protein binds to the UAS, resulting in expression of beta-lactamase. Division Arrested (DA) cells are available in two configurations- an Assay Kit (which includes cells and sufficient substrate to analyze 1 x 384-well plate), and a tube of cells sufficient to analyze 10 x 384-well plates. PPAR gamma-UAS-bla HEK 293H cells are functionally validated for Z' and EC50 concentrations of Rosiglitazone. In addition, PPAR gamma-UAS-bla HEK 293H cells have been tested for assay performance under variable conditions, including DMSO concentration, cell number, and stimulation time.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cell-based beta lactamase reporter gene
PPAR gamma-UAS-bla 293H cells (K1094)

Shipping: Dry Ice
Storage: -80°C (liquid nitrogen), Liquid Nitrogen
Nuclear Receptors
Nuclear Receptors
GeneBLAzer™
HEK 293H Cells
PPARG, PPAR gamma, NR1C3
Dividing Cells
Fluorescence, Fluorescent
Human
Calcitriol
2 x 106 Cells
BLA (Beta-Lactamase)
Dividing Cells
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.