missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneBLAzer™ PTGDR-CRE-bla CHO-K1 Cells

GeneBLAzer™ PTGDR-CRE-bla CHO-K1 cells contain the human Prostanoid (PTGDR) Receptor stably integrated into the CellSensor™ CRE-bla CHO-K1 cell line.

Brand:  Thermo Scientific™ K1739

Product Code. 10647764

  • £10780.00 / Each
Estimated Shipment: 16-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

CellSensor™ CRE-bla CHO-K1 cells (Cat. no. K1535) contain a beta-lactamase reporter gene under control of the Cyclic AMP Response Element (CRE) response element.

The GeneBLAzer™ PTGDR-CRE-bla CHO-K1cells are functionally validated for Z' and EC50 concentrations of Prostaglandin D2. In addition, GeneBLAzer™ PTGDR-CRE-bla CHO-K1 cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary.

The PTGDR receptor also known as DP, is a Gs-coupled receptor which activates adenylate cyclase. PTGDR receptors are located in vascular smooth muscle cells and blood platelets, and are also believed to be expressed in the brain, where they may be involved in the regulation of sleep. In addition, knockout studies in mice suggest that PTGDR may be involved in allergic and asthmatic responses. Potent agonists and antagonists for PTGDR have been identified.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

NM_000953.2
Dividing Cells
Fluorescence
PTGDR
GeneBLAzer™
BLA (Beta-Lactamase)
CHO-K1
GPCR
Cell-based beta lactamase reporter gene
Contents:
K1739 GeneBLAzer™ PTGDR-CRE-bla CHO-K1 cells
(LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol).
Includes:
• GeneBLAzer™ PTGDR-CRE-bla CHO-K1 cells
• GeneBLAzer™ PTGDR-CRE-bla CHO-K1 Assay Protocol
• Certificate of Analysis

Storage:
The GeneBLAzer™ PTGDR-CRE-bla CHO-K1 cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.
FRET
Single Cell Clone Origin
2 x 106 Cells
PTGDR
GeneBLAzer™
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.