missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneBLAzer™ RAR alpha DA Assay Kit

The GeneBLAzer™RAR alpha DA (Division Arrested) and RAR alpha-UAS-bla HEK 293T cells contain the ligand-binding domain (LBD) of the human retinoic acid receptor alpha fused to the DNA-binding domain of GAL4 stably integrated in the GeneBLAzer™UAS-bla HEK293T cell line.

Brand:  Thermo Scientific™ K1387

Product Code. 10276313

  • £946.00 / Each
Estimated Shipment: 03-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

The GeneBLAzer™RAR alpha DA (Division Arrested) and RAR alpha-UAS-bla HEK 293T cells contain the ligand-binding domain (LBD) of the human retinoic acid receptor alpha fused to the DNA-binding domain of GAL4 stably integrated in the GeneBLAzer™UAS-bla HEK293T cell line. GeneBLAzer™UAS-bla HEK 293T cells (Cat. No. K1104) stably express a beta-lactamase reporter gene under the transcriptional control of a 7X Upstream Activator Sequence (UAS). Transcription from the 7X UAS is activated by the binding of the GAL4 transcription factor DNA-binding-domain (DBD). The GAL4-DBD is expressed as a fusion protein with the ligand binding domain (LBD) of RAR alpha. When an agonist binds to the LBD of the GAL4(DBD)-RAR alpha(LBD) fusion protein it translocates to the nucleus where it binds to the 7X UAS inducing transcription of beta-lactamase.

Division Arrested (DA) cells are available in an Assay Kit (which includes cells and sufficient substrate to analyze 1 x 384-well plate). DA cells are irreversibly division arrested using a low-dose treatment of Mitomycin-C, and have no apparent toxicity or change in cellular signal transduction. Both RAR alpha DA cells and RAR alpha-UAS-bla HEK 293T cells have been tested for assay performance using variable assay conditions, including DMSO concentration, cell number, stimulation time, substrate loading time and have been validated for Z' and EC50 concentrations of all-trans retinoic acid. Additional testing data using alternate stimuli are also available.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Tetrac, T4
Cell-based beta lactamase reporter gene
Contains sufficient division-arrested cells and substrate to assay 1 x 384-well plate:
• RAR alpha HEK 293T DA cells (K1387A)
• LiveBLAzer™-FRET B/G Loading Kit, 70 μg

Shipping: Dry Ice
Storage: -80°C (liquid nitrogen)
Fluorescence
RARα
T3 Hormone
1 plate
293T (HEK)
Division-Arrested Cells Assay Kit
GeneBLAzer™
1-850
Division-Arrested Cells
GeneBLAzer™ RAR alpha DA Assay Kit, 1 Plate, ∽4, 800,000 No. of Cell, Liquid, Division-Arrested Cells State, 293T (HEK) Cell Line, Fluorescent Detection Method, Nuclear Receptors Target, BLA (Beta-Lactamase) Reporter, Epithelial Cell Phenotype, Adherent Growth Properties
Cellular imaging, pharma and biopharma, target and lead identification and validation, target-based adme/tox assays
RAR alpha HEK 293T DA cells (K1387A) LiveBLAzer™-FRET B/G loading kit
GeneBLAzer™
BLA (Beta-Lactamase)
RARA, RAR alpha, NR1B1
293T (HEK)
Nuclear Receptors
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.