Learn More
Corning™ General Assay Microplate Lids

Manufactured with rigid polystyrene for optimum visibility and strength. Corning™ General Assay Microplate Lids are available in a variety of styles and types.
Brand: Corning™ 3099
105.69 GBP valid until 2025-04-13
Use promo code "24122" to get your promotional price.
Description

Specifications
Polystyrene | |
50/Cs. | |
Universal Lid | |
Corner Notch |
Sterile | |
25/Pk. | |
96/384-well, Half Area Plate |
Product Suggestions
Customers who viewed this item also viewed
Special Offers
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
Days
13Hours
11Minutes
57Seconds
21New Offer Save Up to 92% -
Corning Matrigel
,
Corning Biocoat
,
Corning CoolProduct
,
Corning Media Tech
,
Corning
Your input is important to us. Please complete this form to provide feedback related to the content on this product.