Learn More
Corning™ General Assay Microplate Lids
Manufactured with rigid polystyrene for optimum visibility and strength. Corning™ General Assay Microplate Lids are available in a variety of styles and types.
Brand:  Corning™ 3095
 View more versions of this product
Product Code. 10402751
Description
Description
  • All lids are made of rigid polystyrene except where indicated.
  • All lids have a corner notch on the A1 corner (except where indicated) to correspond to the corner notches found on all Corning™ microplates.
  • The universal lid without a corner notch (Cat. No. 3098) does not need to be oriented in any particular direction to be placed on Corning microplates. The lid also has a shorter skirt than standard lids.
  • The black universal lid (Cat. No. 3935) is suitable for fluorescent and other light-sensitive assays.
  • The DMSO-resistant cyclic olefin copolymer (COC) lid (Cat. No. 3085) is tinted amber in color for light-sensitive assays and is 100% DMSO-resistant.
  • TRUSTED_SUSTAINABILITY
    Specifications
    Specifications
    Acetate
    100
    Sealing Tape
    Non-sterile
    100
    96-well Plate
    Product Suggestions
    Product Suggestions
    Videos
    SDS
    Documents
    Documents
    Certificates
    View accessory products
    Special Offers
    Special Offers
    Product Content Correction
    Product Title
    Corning™ General Assay Microplate Lids > Acetate sealing tape; Case of 100