Learn More
Thermo Scientific™ GENESYS™ 140/150 Vis/UV-Vis Spectrophotometers
Get the most for your QA/QC or teaching lab with a rugged, network ready spectrophotometer designed for repetitive measurements and ease of use.
£3687.30 - £6385.00
Products 3
Product Code Brand Description Spectral Bandwidth Wavelength Range Price Quantity & Availability  
Product Code Brand Description Spectral Bandwidth Wavelength Range Price Quantity & Availability  
15820002
View Documents
Thermo Scientific™
840-308400
GENESYS 140 Visible Spectrophotometer 5 nm 325 to 1100 nm N/A N/A N/A
15830002
View Documents Promotion Details
Thermo Scientific™
840-300300
GENESYS 150 UV-Vis Spectrophotometer with EU Power Cable 2 nm 190 to 1100 nm
£6385.00
Each
15840002
View Documents Promotion Details
Thermo Scientific™
840-300400
GENESYS 150 UV-Vis Spectrophotometer with UK Power Cable 2 nm 190 to 1100 nm
£6370.00
Each
Description
Show More
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Thermo Scientific™ GENESYS™ 140/150 Vis/UV-Vis Spectrophotometers