Learn More
Thermo Scientific™ GENESYS™ 30 Visible Spectrophotometer Accessories
Get all of the tools you need to facilitate your analytical process with accessories for the Thermo Scientific™ GENESYS™ 30 Visible Spectrophotometer.
Brand: Thermo Scientific™ 840-278700
133.00 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Description
Get all of the tools you need to facilitate your analytical process with accessories for the Thermo Scientific™ GENESYS™ 30 Visible Spectrophotometer.
Pre-aligned, pre-assembled replacement lampCustomer replaceable - slides into place
Mouse-hole replacement access. Cover removal not required.
Specifications
Pre-aligned, pre-assembled replacement Tungsten-Halogen lamp. Mouse-hole replacement access. Cover removal not required. | |
Tungsten Halogen Lamp | |
GENESYS 30 Visible Spectrophotometer | |
1 pre-aligned lamp assembly |
Product Suggestions
Customers who viewed this item also viewed.