missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Gentamicin (10 mg/mL)

Gentamicin (10 mg/mL)
£151.00 - £2180.00
Specifications
Certifications/Compliance | ISO 13485, cGMP, FDA |
---|---|
Content And Storage | 15 to 30°C |
Concentration | 10 mg/mL |
Form | Liquid |
Product Type | Antibiotic |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11500506
![]() |
Gibco™
15710049 |
100 mL |
£151.00
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11510506
![]() |
Gibco™
15710080 |
20 x 100mL |
£2180.00
Pack of 20 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Gentamicin sulfate is a water-soluble antibiotic drug originally purified from the fungus Micromonospora purpurea. Gentamicin acts by binding to the 30S subunit of the bacterial ribosome leading to inhibition protein synthesis and death in susceptible bacteria. Gibco™ Gentamicin is effective against a wide variety of gram-positive and gram-negative bacteria, and is used for the prevention of bacterial contamination on cell cultures. The recommended working concentration ranges from 0.5 to 50μg/ml. We offer a variety of antibiotics and antimycotics for cell culture applications.Product Use
For Research Use Only: Not intended for animal or human diagnostic or therapeutic use.
Dual-Site cGMP Manufacturing
Gibco™ Gentamicin is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ Gentamicin product made in our Grand Island facility (15710-072). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
Specifications
ISO 13485, cGMP, FDA | |
10 mg/mL | |
Antibiotic |
15 to 30°C | |
Liquid | |
Mammalian cell culture, prevention of cell culture contamination |
Safety and Handling
missing translation for 'shelfLife' : 24 Months
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- GENTAMICIN (10MG/ML)
Signal Word
- Danger
Hazard Category
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : gentamicin sulphate
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Gibco™ Gentamicin (10 mg/mL)