missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Grade 907 Germination and Seed Test Papers, Sheet
For germination assays of seed specimens according to various methods such as Neeb and Jacobsen.
£21.60 - £41.75
Specifications
Basis Weight | 160 g/m2 |
---|---|
Color | White |
Material | Paper |
For Use With (Application) | Germination assays of seed specimen |
No. per Pack | 100 |
Product Code | Brand | Width (Metric) | No. per Pack | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Width (Metric) | No. per Pack | Price | Quantity & Availability | ||||
11709915
|
Fisherbrand™
|
170 x 110mm | 100 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
11799905
|
Fisherbrand™
|
155 x 175mm | 100 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Wetstrength paper 160g/m2
- Absorbent
- Free from chlorine
- pH must be between 6.0 and 7.5
- For small seeds such as grasses and flowers
- White in color
Specifications
160 g/m2 | |
Paper | |
100 | |
0.38 mm |
White | |
Germination assays of seed specimen | |
6 to 7.5 | |
Germination and Seed Test Paper |