Learn More
Gibco™ Advanced DMEM

Advanced DMEM (Dulbecco's Modified Eagle Medium) is a widely used basal medium that allows the culture of mammalian cells with reduced Fetal Bovine Serum (FBS) supplementation.
£25.45 - £224.00
Specifications
Cell Line | MDBK, HepG2, COS-7, A549, MDCK, WI-38, and Vero |
---|---|
Classification | Animal Origin |
Concentration | 1 X |
Form | Liquid |
Product Type | DMEM (Dulbecco's Modified Eagle Medium) |
Requires: Supplementation
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11510436
![]() |
Gibco™
12491015 |
500 mL |
£25.45
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11520436
![]() |
Gibco™
12491023 |
10 x 500 mL |
£224.00
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Advanced DMEM (Dulbecco's Modified Eagle Medium) is a widely used basal medium that allows the culture of mammalian cells with reduced Fetal Bovine Serum (FBS) supplementation. Compared to classic DMEM, serum supplementation can be reduced by 50–90% with no change in growth rate or morphology. Cells successfully cultured in Advanced DMEM, with no adaptation, include MDBK, HepG2, COS-7, A549, MDCK, WI-38, and Vero.
This Advanced DMEM is manufactured as follows:
With: High Glucose, Non-essential Amino Acids, Sodium Pyruvate, Phenol Red
Without: L-glutamine
The complete formulation is available.
Using Advanced DMEM
Advanced DMEM is unique from other media due to the addition of the following ingredients to allow for serum reduction: ethanolamine, glutathione, ascorbic acid, insulin, transferrin, AlbuMAX™ I lipid-rich bovine serum albumin for cell culture, and the trace elements sodium selenite, ammonium metavanadate, cupric sulfate, and manganous chloride. Advanced DMEM requires supplementation with 1–5% Fetal Bovine Serum and 4 mM L-glutamine or GlutaMAX™ supplement. Many cell lines do not require adaptation to this media. The FBS concentration must be optimized for each cell line to obtain maximum serum reduction. Advanced DMEM uses a sodium bicarbonate buffer system (3.7 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH.
Order Info
Shipping Condition: Room Temperature
Specifications
MDBK, HepG2, COS-7, A549, MDCK, WI-38, and Vero | |
1 X | |
DMEM (Dulbecco's Modified Eagle Medium) | |
Sterile-filtered | |
High Glucose, Phenol Red, Sodium Pyruvate, Non Essential Amino Acids (NEAA) | |
cGMP-compliant under the ISO 13485 standard | |
Room Temperature |
Animal Origin | |
Liquid | |
Reduced Serum | |
Sterile-filtered | |
No Glutamine, No HEPES | |
Gibco™ | |
Storage conditions: 2-8° C. Protect from light Shipping conditions: Ambient Shelf life: 12 months from date of manufacture |
Safety and Handling
missing translation for 'shelfLife' : 12 Months From Date of Manufacture
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.