Learn More
Gibco™ Advanced DMEM/F-12
Advanced DMEM/F-12 (Dulbecco's Modified Eagle Medium/Ham's F-12) is a widely used basal medium that allows the culture of mammalian cells with reduced Fetal Bovine Serum (FBS) supplementation.
£29.12 - £255.00
Specifications
Content And Storage | Storage conditions: 2-8° C. Protect from light Shipping conditions: Ambient Shelf life: 12 months from date of manufacture |
---|---|
Cell Line | MRC-5, SP2, Vero, WI-38 and Jurkat |
Classification | Animal Origin |
Concentration | 1 X |
Form | Liquid |
Requires: Supplementation
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11540446
|
Gibco™
12634010 |
500 mL |
£29.12
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11550446
|
Gibco™
12634028 |
10 x 500 mL |
£255.00
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Advanced DMEM/F-12 (Dulbecco's Modified Eagle Medium/Ham's F-12) is a widely used basal medium that allows the culture of mammalian cells with reduced Fetal Bovine Serum (FBS) supplementation. Compared to classic DMEM/F-12, serum supplementation can be reduced by 50–90% with no change in growth rate or morphology. Cells successfully cultured in Advanced DMEM/F-12, with no adaptation, include MRC-5, SP2, Vero, WI-38 and Jurkat.
This Advanced DMEM/F-12 is manufactured as follows:
With: Glucose, Non-essential Amino Acids, Sodium Pyruvate, Phenol Red
Without: L-glutamine, HEPES
The complete formulation is available.
Using Advanced DMEM/F-12
Advanced DMEM/F-12 is unique from other media due to addition of the following ingredients to allow for serum reduction: ethanolamine, glutathione, ascorbic acid, insulin, transferrin, AlbuMAX™ II lipid-rich bovine serum albumin for cell culture, and the trace elements sodium selenite, ammonium metavanadate, cupric sulfate, and manganous chloride. Advanced DMEM/F-12 requires supplementation with 1–5% Fetal Bovine Serum and 4 mM L-glutamine or GlutaMAX™ supplement. Many cell lines do not require adaptation to this media. The FBS concentration must be optimized for each cell line to obtain maximum serum reduction. Advanced Advanced DMEM/F-12 uses a sodium bicarbonate buffer system (3.7 g/L) and therefore requires a 5–10% CO2 environment to maintain physiological pH.
Order Info
Shipping Condition: Room Temperature
Specifications
Storage conditions: 2-8° C. Protect from light Shipping conditions: Ambient Shelf life: 12 months from date of manufacture |
|
Animal Origin | |
Liquid | |
Reduced Serum | |
High Glucose, Phenol Red, Sodium Pyruvate, Non Essential Amino Acids (NEAA) | |
Sustainable packaging | |
Gibco™ |
MRC-5, SP2, Vero, WI-38 and Jurkat | |
1 X | |
DMEM (Dulbecco's Modified Eagle Medium)/F-12 | |
Sterile-filtered | |
No Glutamine, No HEPES | |
cGMP-compliant under the ISO 13485 standard | |
Room Temperature |
Safety and Handling
missing translation for 'shelfLife' : 12 Months From Date of Manufacture
- Methyl benzoate
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H302-Harmful if swallowed.
Precautionary Statement
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
Supplemental information
- MIXTURE LIST-Contains : Methyl benzoate
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.