Learn More
Gibco™ DMEM, high glucose, NEAA, no glutamine

Dulbecco's Modified Eagle Medium (DMEM) is a widely used basal medium for supporting the growth of many different mammalian cells.
Brand: Gibco™ 10938025
12.78 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Description
Dulbecco's Modified Eagle Medium (DMEM) is a widely used basal medium for supporting the growth of many different mammalian cells. Cells successfully cultured in DMEM include primary fibroblasts, neurons, glial cells, HUVECs and smooth muscle cells, as well as cell lines such as HeLa, 293, Cos-7, and PC-12. We offer a variety of Gibco™ DMEM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.
This DMEM is modified as follows:
With: High Glucose, Non-essential Amino Acids, Phenol Red
Without: L-glutamine, Sodium Pyruvate, HEPES
The complete formulation is available.
DMEM is unique from other media as it contains 4 times the concentration of amino acids and vitamins than the original Eagle's Minimal Essential Medium. DMEM was originally formulated with low glucose (1 g/L) and sodium pyruvate, but is often used with higher glucose levels, with or without sodium pyruvate.
Product Use
For Research Use Only: Not intended for animal or human diagnostic or therapeutic use.
DMEM contains no proteins, lipids, or growth factors. Therefore, DMEM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). DMEM uses a sodium bicarbonate buffer system (3.7 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.
Order Info
Shipping Condition: Room Temperature
Compliance
Gibco DMEM is manufactured at a cGMP compliant facility located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard.

Specifications
HeLa, 293, Cos-7, and PC-12 | |
Animal Origin-free | |
Liquid | |
Standard Serum Supplementation | |
High Glucose, Phenol Red, Non Essential Amino Acids (NEAA), Sodium Bicarbonate | |
cGMP-compliant under the ISO 13485 standard | |
500 mL | |
310 - 360 mOsm/kg | |
CO2 |
Primary Fibroblasts, Neurons, Glial Cells, HUVECs, Smooth Muscle Cells | |
1 X | |
DMEM (Dulbecco's Modified Eagle Medium) | |
Sterile-filtered | |
No Glutamine, No HEPES, No Sodium Pyruvate | |
Gibco™ | |
Room Temperature | |
Storage conditions: 2-8° C. Protect from light. Shipping conditions: Ambient. Shelf life: 12 months from date of manufacture. |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2656891 | Certificate of Analysis | 29/01/2025 | 10938025 |
2842977 | Certificate of Analysis | 26/01/2025 | 10938025 |
2916972 | Certificate of Analysis | 25/01/2025 | 10938025 |
3042503 | Certificate of Analysis | 25/01/2025 | 10938025 |
A0428823 | Certificate of Analysis | 22/04/2024 | 109385000,10938025,109381000,109380050 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.