missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ DMEM, powder, high glucose, pyruvate

DMEM (Dulbecco's Modified Eagle Medium) is a widely used basal medium for supporting the growth of many different mammalian cells.

Brand:  Gibco™ 12800017

 View more versions of this product

Product Code. 11588826

  • £39.99 / 10 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 

Requires : Supplementation
This item is not returnable. View return policy

Description

Description

DMEM (Dulbecco's Modified Eagle Medium) is a widely used basal medium for supporting the growth of many different mammalian cells. Cells successfully cultured in DMEM include primary fibroblasts, neurons, glial cells, HUVECs, and smooth muscle cells, as well as cell lines such as HeLa, 293, Cos-7, and PC-12. We offer a variety of DMEM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

This DMEM is modified as follows:
With: High Glucose, L-glutamine, Phenol Red, Sodium Pyruvate
Without: HEPES, Sodium Bicarbonate

The complete formulation is available.

Using DMEM
DMEM is unique from other media as it contains 4 times the concentration of amino acids and vitamins than the original Eagle's Minimal Essential Medium. DMEM was originally formulated with low glucose (1 g/L) and sodium pyruvate, but is often used with higher glucose levels, with or without sodium pyruvate. DMEM contains no proteins, lipids, or growth factors. Therefore, DMEM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). DMEM uses a sodium bicarbonate buffer system (3.7 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH. Powder forms of Gibco™ cell culture media require sodium bicarbonate supplementation, pH adjustment, and filtration at the time of preparation (see protocol for details).

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

HeLa, 293, Cos-7, and PC-12
Animal Origin-free
DMEM (Dulbecco's Modified Eagle Medium)
Sterile-filtered
No HEPES, No Sodium Bicarbonate
Gibco™
Room Temperature
Primary Fibroblasts, Neurons, Glial Cells, HUVECs, Smooth Muscle Cells
Powder
Standard Serum Supplementation
High Glucose, Glutamine, Phenol Red, Sodium Pyruvate
cGMP-compliant under the ISO 13485 standard
10 x 1 L
Storage conditions: 2-8° C
Shipping conditions: Ambient
Shelf life: 36 months from date of manufacture
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2782383 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number12800017,12800082
Lot Number2788681 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number12800017,12800082
Lot Number2725161 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number12800017,12800082
Lot Number2782384 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number12800017,12800082
Lot Number2782431 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number12800017,12800082
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ DMEM, powder, high glucose, pyruvate > 10 x 1 L

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.