missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Fetal Bovine Serum, certified, United States

Important Update: We've recently introduced Premium Plus Fetal Bovine Serum (FBS) specification-based products. These improved products are alternatives to our Certified US origin, New Zealand origin, and Qualified Australia origin SKUs. Product stability, performance, manufacturing, storage, and transport processes are unchanged.

Brand:  Gibco™ A3160402

Additional Details : Weight : 0.04540kg

 View more versions of this product

Product Code. 15565309

  • £590.00 / Pack of 10
In Stock
to see stock.

Description

Description

Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements

Gibco Sera: Trusted Quality for Consistent Performance

Sera Category: Premium (Performance Plus)

  • Use for a broad range of cell types, especially sensitive cell lines
  • Our most characterized sera with the lowest endotoxin and hemoglobin levels
  • Endotoxin level: ≤5 EU/mL
  • Hemoglobin level: ≤15 mg/dL
  • Origin: United States

Gibco Serum Delivers

  • ISO 13485 certified, processed in FDA registered facilities
  • Triple filtered at 0.1 μm
  • Gibco bottle is easier to use in the hood, reduces the risk of contamination and helps you perform cell culture more consistently
  • Peel-off sticker on label provides handy reference for lot number and expiration date recording in lab notebook
Specifications

Specifications

Storage conditions: ≤-10°C
Shipping conditions: Frozen
Fetal
United States
One Shot
10 x 50 mL
Sterile
≤5 EU/mL
≤15 mg/dL
One Shot™
Fetal Bovine Serum, certified, One Shot™ format, United States
Bovine
Liquid
Fetal Bovine Serum
Standard (Sterile-filtered)
Triple-filtered, 0.1 μm
Fewer resources used and less waste
Certified
Frozen
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.