Learn More
Gibco™ Fetal Bovine Serum, certified, United States
Important Update: We've recently introduced Premium Plus Fetal Bovine Serum (FBS) specification-based products. These improved products are alternatives to our Certified US origin, New Zealand origin, and Qualified Australia origin SKUs. Product stability, performance, manufacturing, storage, and transport processes are unchanged.
£63.50 - £590.00
Specifications
Content And Storage | Storage conditions: ≤-10°C Shipping conditions: Frozen |
---|---|
Age | Fetal |
Country of Origin | United States |
Form | Liquid |
Product Type | Fetal Bovine Serum |
Product Code | Brand | Quantity | Format | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Format | Price | Quantity & Availability | ||||
15555309
|
Gibco™
A3160401 |
50 mL | One Shot |
£63.50
50mL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15565309
|
Gibco™
A3160402 |
10 x 50 mL | One Shot |
£590.00
Pack of 10 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11570506
|
Gibco™
16000044 |
500 mL | Bottle |
£494.00
500mL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11560506
|
Gibco™
16000036 |
100 mL | Bottle |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements
Gibco Sera: Trusted Quality for Consistent Performance
Sera Category: Premium (Performance Plus)
- Use for a broad range of cell types, especially sensitive cell lines
- Our most characterized sera with the lowest endotoxin and hemoglobin levels
- Endotoxin level: ≤5 EU/mL
- Hemoglobin level: ≤15 mg/dL
- Origin: United States
Gibco Serum Delivers
- ISO 13485 certified, processed in FDA registered facilities
- Triple filtered at 0.1 μm
- Gibco bottle is easier to use in the hood, reduces the risk of contamination and helps you perform cell culture more consistently
- Peel-off sticker on label provides handy reference for lot number and expiration date recording in lab notebook
Order Info
Shipping Condition: Dry Ice
Specifications
Storage conditions: ≤-10°C Shipping conditions: Frozen |
|
United States | |
Fetal Bovine Serum | |
Sterile | |
≤5 EU/mL | |
Certified |
Fetal | |
Liquid | |
Standard (Sterile-filtered) | |
Triple-filtered, 0.1 μm | |
≤15 mg/dL | |
Frozen |
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.