Learn More
Gibco™ Fetal Bovine Serum, qualified, New Zealand

Important Update: We've recently introduced Premium Plus Fetal Bovine Serum (FBS) specification-based products. These improved products are alternatives to our Certified US origin, New Zealand origin, and Qualified Australia origin SKUs. Product stability, performance, manufacturing, storage, and transport processes are unchanged.
£105.00 - £978.00
Specifications
Content And Storage | Storage conditions: ≤-10°C Shipping conditions: Frozen |
---|---|
Age | Fetal |
Country of Origin | New Zealand |
Form | Liquid |
Product Type | Fetal Bovine Serum |
Product Code | Brand | Quantity | Format | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Format | Price | Quantity & Availability | |||||
15505319
![]() |
Gibco™
A3160901 |
50 mL | One Shot | N/A | ||||||
15515319
![]() |
Gibco™
A3160902 |
10 x 50 mL | One Shot | N/A | ||||||
11548836
![]() |
Gibco™
10091148 |
500 mL | Bottle | N/A | ||||||
Description
Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements
- Sera Category: Premium (Secure)
- Origin: New Zealand
- Endotoxin level: ≤5 EU/mL
- Hemoglobin level: ≤30 mg/dL (levels routinely ≤25 mg/dL)
Order Info
Shipping Condition: Dry Ice
Specifications
Storage conditions: ≤-10°C Shipping conditions: Frozen |
|
New Zealand | |
Fetal Bovine Serum | |
Sterile | |
≤30 mg/dL |
Fetal | |
Liquid | |
Standard (Sterile-filtered) | |
≤5 EU/mL | |
Frozen |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.