missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Fetal Bovine Serum, qualified, New Zealand
GREENER_CHOICE

Important Update: We've recently introduced Premium Plus Fetal Bovine Serum (FBS) specification-based products. These improved products are alternatives to our Certified US origin, New Zealand origin, and Qualified Australia origin SKUs. Product stability, performance, manufacturing, storage, and transport processes are unchanged.

Brand:  Gibco™ A3160902

 View more versions of this product

Product Code. 15515319

This item is not returnable. View return policy

Description

Description

Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements

  • Sera Category: Premium (Secure)
  • Origin: New Zealand
  • Endotoxin level: ≤5 EU/mL
  • Hemoglobin level: ≤30 mg/dL (levels routinely ≤25 mg/dL)

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Storage conditions: ≤-10°C
Shipping conditions: Frozen
Fetal
New Zealand
One Shot
10 x 50 mL
Sterile
≤30 mg/dL
One Shot™
Fetal Bovine Serum, qualified, One Shot™ format, New Zealand
Bovine
Liquid
Fetal Bovine Serum
Standard (Sterile-filtered)
≤5 EU/mL
Qualified
Frozen
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ Fetal Bovine Serum, qualified, New Zealand > One Shot™, 10 x 50 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.