missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Hybridoma-SFM
GREENER_CHOICE

Specifically developed to support the growth of hybridomas for antibody production

Brand:  Gibco™ 12045076

86.67 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 10749233

  • £107.00 / 1 L

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

  • Hybridoma-SFM is a serum-free, very low-protein (<20 μg/mL) medium specifically developed to support the growth of hybridomas for antibody production
  • Suitable for the culture of recombinant myeloma lines as well as traditional hybridomas, Hybridoma-SFM provides high cell densities and IgG production
  • Cholesterol-dependent cultures such as NS0 require the addition of 250X Cholesterol Lipid Concentrate
  • Hybridoma-SFM is a complete medium that is ready to use, no supplementation required
  • Store at 2°C to 8°C, protected from light

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Storage conditions: 2-8°C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Low Protein, Serum-free
Serum Free Medium (SFM)
Glutamine, Phenol Red
1000 mL
Hybridomas
Liquid
Serum-free
Gibco™
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.