missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Hybridoma-SFM

Specifically developed to support the growth of hybridomas for antibody production
£74.25 - £117.00
Specifications
Content And Storage | Storage conditions: 2-8°C. Protect from light Shipping conditions: Ambient Shelf life: 12 months from date of manufacture |
---|---|
Cell Type | Hybridomas |
Classification | Low Protein, Serum-free |
Form | Liquid |
Product Type | Serum Free Medium (SFM) |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11570416
![]() |
Gibco™
12045084 |
500 mL |
£74.25
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10749233
![]() |
Gibco™
12045076 |
1000 mL |
£117.00
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Hybridoma-SFM is a serum-free, very low-protein (<20 μg/mL) medium specifically developed to support the growth of hybridomas for antibody production
- Suitable for the culture of recombinant myeloma lines as well as traditional hybridomas, Hybridoma-SFM provides high cell densities and IgG production
- Cholesterol-dependent cultures such as NS0 require the addition of 250X Cholesterol Lipid Concentrate
- Hybridoma-SFM is a complete medium that is ready to use, no supplementation required
- Store at 2°C to 8°C, protected from light
Order Info
Shipping Condition: Room temperature
Specifications
Storage conditions: 2-8°C. Protect from light Shipping conditions: Ambient Shelf life: 12 months from date of manufacture |
|
Low Protein, Serum-free | |
Serum Free Medium (SFM) | |
Glutamine, Phenol Red | |
Room Temperature |
Hybridomas | |
Liquid | |
Serum-free | |
Gibco™ |
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Gibco™ Hybridoma-SFM